Learn More
Meridian Life Science Taq HS DNA Polymerase, 100UL
SDP
Catalog No. 5000856485
Encompass_Preferred
Taq HS DNA Polymerase is an antibody-mediated hot-start DNA polymerase provided with an optimized buffer system containing dNTP, MgCl2, stabilizers, and PCR enhancers at optimal concentrations which eliminates the need for optimization. The DNA polymerase has been developed to give more robust amplification than other polymerases allowing it to perform well with challenging templates in the presence of PCR inhibitors, making it ideal for fast PCR reactions across a range of templates, without compromising specificity or yield.
Catalog No. 50-008-56485 Supplier Meridian Life Science Supplier No. MDX00801
Taq HS DNA Polymerase is an antibody-mediated hot-start DNA polymerase provided with an optimized buffer system containing dNTP, MgCl2, stabilizers, and PCR enhancers at optimal concentrations which eliminates the need for optimization. The DNA polymerase has been developed to give more robust amplification than other polymerases allowing it to perform well with challenging templates in the presence of PCR inhibitors, making it ideal for fast PCR reactions across a range of templates, without compromising specificity or yield.