Learn More
New England Biolabs, Inc. TaqI Methyltransferase – 1000 units
SDP
Catalog No. 50811638
Encompass
TaqI Methyltransferase modifies the adenine residue (N6) of the sequence TCGA.
Catalog No. 50-811-638 Supplier New England Biolabs, Inc. Supplier No. M0219S
TaqI Methyltransferase modifies the adenine residue (N6) of the sequence TCGA.