Learn More
Takara Bio TAQ (MG2+ FREE BUFFER) 250U
SDP
Catalog No. 50443949
Encompass
PCR Reagent; Takara; Taq DNA Polymerase (dNTPs, magnesium-free buffer, and a separate tube of 25mM MgCl2 included); 250U
Catalog No. 50-443-949 Supplier Takara Bio Supplier No. R001AM
PCR Reagent; Takara; Taq DNA Polymerase (dNTPs, magnesium-free buffer, and a separate tube of 25mM MgCl2 included); 250U