Learn More
Meridian Life Science Taq PCR Buffer 5x, 100ML
SDP
Catalog No. 5000856278
Encompass_Preferred
Taq PCR Buffer is a combination of the latest advances in buffer chemistry together with enhancers, stabilizers, dNTPs, and MgCl2 at optimal concentrations. Taq PCR Buffer has been designed to deliver very fast, high yield PCR amplification over a wide range of PCR templates and has been developed to give very robust amplification allowing it to perform well with challenging templates in the presence of PCR inhibitors.
Catalog No. 50-008-56278 Supplier Meridian Life Science Supplier No. MDX002100
Taq PCR Buffer is a combination of the latest advances in buffer chemistry together with enhancers, stabilizers, dNTPs, and MgCl2 at optimal concentrations. Taq PCR Buffer has been designed to deliver very fast, high yield PCR amplification over a wide range of PCR templates and has been developed to give very robust amplification allowing it to perform well with challenging templates in the presence of PCR inhibitors.