Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioland Scientific LLC 2x Taq PCR Premix, ready-to-use solution, 1000 units, maximum amplicon size 4 kb (genomic), 5 kb (lambda), tracking dyes blue and yellow
SDP

Catalog No. NC2753303
Encompass

This 2x Taq PCR Premix is a ready-to-use, optimized solution designed for high-throughput and routine PCR applications. It contains Taq DNA polymerase and necessary components for efficient amplification. Ideal for sub-cloning, genotyping, and more, this premix allows for direct loading onto agarose gels due to included tracking dyes. The formulation supports amplification of DNA fragments from both genomic and lambda DNA templates, ensuring reliable performance.

  • Formulation: 2x concentrated premix
  • Enzyme type: Taq DNA polymerase, recombinant
  • Maximum amplicon size (genomic DNA): 4 kb
  • Maximum amplicon size (lambda DNA): 5 kb
  • Storage temperature (long-term): -20 °C
  • Storage temperature (short-term): 4 °C (stable up to 6 weeks)
  • Freeze-thaw stability: Up to 10 cycles
  • Applications: PCR, primer extension, microarray analysis, high-throughput PCR
  • Format: Ready-to-use solution

This item is not returnable. View return policy
Catalog No. NC2753303 Supplier Bioland Scientific LLC Supplier No. TP0102
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 2x Taq PCR Premix is a ready-to-use, optimized solution designed for high-throughput and routine PCR applications. It contains Taq DNA polymerase and necessary components for efficient amplification. Ideal for sub-cloning, genotyping, and more, this premix allows for direct loading onto agarose gels due to included tracking dyes. The formulation supports amplification of DNA fragments from both genomic and lambda DNA templates, ensuring reliable performance.

  • Formulation: 2x concentrated premix
  • Enzyme type: Taq DNA polymerase, recombinant
  • Maximum amplicon size (genomic DNA): 4 kb
  • Maximum amplicon size (lambda DNA): 5 kb
  • Storage temperature (long-term): -20 °C
  • Storage temperature (short-term): 4 °C (stable up to 6 weeks)
  • Freeze-thaw stability: Up to 10 cycles
  • Applications: PCR, primer extension, microarray analysis, high-throughput PCR
  • Format: Ready-to-use solution

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.