Learn More
Meridian Life Science Taq PCR Red Buffer 5x, 100ML
SDP
Supplier:  Meridian Life Science MDX002100A
Encompass_Preferred
A novel buffer system containing a red gel loading dye, together with enhancers, stabilizers, dNTPs, and MgCl2 at optimal concentrations. Taq PCR Buffer has been designed to deliver very fast, high yield PCR amplification over a wide range of PCR templates and has been developed to give very robust amplification allowing it to perform well with challenging templates in the presence of PCR inhibitors.
Catalog No. 50-008-56279