Learn More
LAMDA BIOTECH INC Taq Plus 2x Master Mix 25ml
Catalog No. NC3726060
Encompass_Preferred
Taq Plus™ 2X PCR MasterMix is an advanced ready-to-use formulation combining enhanced Taq DNA polymerase with high-fidelity polymerase for improved PCR performance. This bulk format is ideal for high-throughput applications and large-scale experiments.
Catalog No. NC3726060 Supplier LAMDA BIOTECH INC Supplier No. D124R25
Taq Plus™ 2X PCR MasterMix is an advanced ready-to-use formulation combining enhanced Taq DNA polymerase with high-fidelity polymerase for improved PCR performance. This bulk format is ideal for high-throughput applications and large-scale experiments.