Learn More
Takara Bio TAQ POLYM HOT-START 1000U
SDP
Catalog No. 50443958
Encompass
PCR Reagents; Takara; Hot-Start Taq DNA Polymerase; 1000U (4 x 250U); reduces background, increased specificity, permits room-temperature reaction assembly
Catalog No. 50-443-958 Supplier Takara Bio Supplier No. R007B
PCR Reagents; Takara; Hot-Start Taq DNA Polymerase; 1000U (4 x 250U); reduces background, increased specificity, permits room-temperature reaction assembly