Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free, 5,000U
Catalog No. 501649564
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9564 Supplier As One International Inc Supplier No. AO100007
Taq DNA Polymerase 5 U/ul glycerol free. Glycerol free is ideal for automation and free drying applications