Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, 10,000U
Catalog No. 501649570
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer. Glycerol free is ideal for automation and free drying applications.
Catalog No. 50-164-9570 Supplier As One International Inc Supplier No. AO101108
Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer. Glycerol free is ideal for automation and free drying applications.