Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, 2,500U
Catalog No. 501649608
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9608 Supplier As One International Inc Supplier No. AO103106
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer. Glycerol free is ideal for automation and free drying applications