Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, 5,000U
Supplier:  As One International Inc AO103107
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9609