Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Magnesium free, 10,000U
Catalog No. 501649575
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Mg2+ free. Glycerol free is ideal for automation and free drying applications.
Catalog No. 50-164-9575 Supplier As One International Inc Supplier No. AO101208
Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Mg2+ free. Glycerol free is ideal for automation and free drying applications.