Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free, 10x Ammonium Buffer and 10x Standard Buffer both Magnesium and detergent free, 5,000U
Catalog No. 501655579
Encompass_Preferred
Taq DNA Polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa. Taq DNA Polymerase has both a 5' to 3' DNA polymerase and a 5' to 3' exonuclease activity. The enzyme lacks a 3' to 5' exonuclease activity (no proofreading ability). Taq DNA Polymerase leaves an A prime overhang, which makes the enzyme ideal for TA cloning. Glycerol free Taq is ideal for automation and free drying applications. Included with Taq are Tween/Triton/magnesium free 10x Ammonium Buffer and 10x Standard Buffer.
Catalog No. 50-165-5579 Supplier As One International Inc Supplier No. AO104507
Taq DNA Polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa. Taq DNA Polymerase has both a 5' to 3' DNA polymerase and a 5' to 3' exonuclease activity. The enzyme lacks a 3' to 5' exonuclease activity (no proofreading ability). Taq DNA Polymerase leaves an A prime overhang, which makes the enzyme ideal for TA cloning. Glycerol free Taq is ideal for automation and free drying applications. Included with Taq are Tween/Triton/magnesium free 10x Ammonium Buffer and 10x Standard Buffer.