Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer, Magnesium free and Triton free, 10,000U
Catalog No. 501649605
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer, Mg2+ free and Triton free. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9605 Supplier As One International Inc Supplier No. AO102508
Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer, Mg2+ free and Triton free. Glycerol free is ideal for automation and free drying applications