Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Magnesium free and Tween free, 2,500U
Catalog No. 501649623
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Mg2+ free and Tween free. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9623 Supplier As One International Inc Supplier No. AO103506
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Mg2+ free and Tween free. Glycerol free is ideal for automation and free drying applications