Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer, 2,500U
Catalog No. 501649588
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer. Glycerol free is ideal for automation and free drying applications.
Catalog No. 50-164-9588 Supplier As One International Inc Supplier No. AO102106
Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer. Glycerol free is ideal for automation and free drying applications.