Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer, 5,000U
Catalog No. 501649589
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer. Glycerol free is ideal for automation and free drying applications.
Catalog No. 50-164-9589 Supplier As One International Inc Supplier No. AO102107
Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer. Glycerol free is ideal for automation and free drying applications.