Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer, Triton free, 10,000U
Catalog No. 501649600
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer, Triton free. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9600 Supplier As One International Inc Supplier No. AO102408
Taq DNA Polymerase 5 U/ul glycerol free 10x Standard Buffer, Triton free. Glycerol free is ideal for automation and free drying applications