Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Tween free, 10,000U
Catalog No. 501649580
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Tween free. Glycerol free is ideal for automation and free drying applications.
Catalog No. 50-164-9580 Supplier As One International Inc Supplier No. AO101408
Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Tween free. Glycerol free is ideal for automation and free drying applications.