Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Tween free, 500U
Catalog No. 501649576
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Tween free. Glycerol free is ideal for automation and free drying applications.
Catalog No. 50-164-9576 Supplier As One International Inc Supplier No. AO101403
Taq DNA Polymerase 5 U/ul glycerol free 10x Ammonium Buffer, Tween free. Glycerol free is ideal for automation and free drying applications.