Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free, 1,000U
Catalog No. 501649617
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9617 Supplier As One International Inc Supplier No. AO103404
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free. Glycerol free is ideal for automation and free drying applications