Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free, 5,000U
Catalog No. 501649619
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9619 Supplier As One International Inc Supplier No. AO103407
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free. Glycerol free is ideal for automation and free drying applications