Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free, 10,000U
Catalog No. 501649620
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9620 Supplier As One International Inc Supplier No. AO103408
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free. Glycerol free is ideal for automation and free drying applications