Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free, 500U
Catalog No. 501649616
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free. Glycerol free is ideal for automation and free drying applications
Catalog No. 50-164-9616 Supplier As One International Inc Supplier No. AO103403
Taq DNA Polymerase 5 U/ul glycerol free 10x Combination Buffer, Tween free. Glycerol free is ideal for automation and free drying applications