Learn More
As One International Inc Taq DNA Polymerase 5 U/ul glycerol free, 500U
Catalog No. 501649561
Encompass_Preferred
Taq DNA Polymerase 5 U/ul glycerol free. Glycerol free is ideal for automation and free drying applications.
Catalog No. 50-164-9561 Supplier As One International Inc Supplier No. AO100003
Taq DNA Polymerase 5 U/ul glycerol free. Glycerol free is ideal for automation and free drying applications.