Learn More
As One International Inc Taq DNA Polymerase 5 U/ul, 10x Ammonium Buffer and 10x Combination buffer both Magnesium and detergent free , 1,000U
Catalog No. 501655702
Encompass_Preferred
AS ONE's Taq DNA Polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa.Taq DNA Polymerase has both a 5' to 3' DNA polymerase and a 5' to 3' exonuclease activity. The enzyme lacks a 3' to 5' exonuclease activity (no proofreading ability). Taq DNA Polymerase leaves an A prime overhang, which makes the enzyme ideal for TA cloning. Taq Polymerase includes Tween/Triton/Magnesium free 10x Ammonium Buffer and 10x Combination Buffer.
Catalog No. 50-165-5702 Supplier As One International Inc Supplier No. AO115504
AS ONE's Taq DNA Polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa.Taq DNA Polymerase has both a 5' to 3' DNA polymerase and a 5' to 3' exonuclease activity. The enzyme lacks a 3' to 5' exonuclease activity (no proofreading ability). Taq DNA Polymerase leaves an A prime overhang, which makes the enzyme ideal for TA cloning. Taq Polymerase includes Tween/Triton/Magnesium free 10x Ammonium Buffer and 10x Combination Buffer.