Learn More
As One International Inc Taq DNA Polymerase 5 U/ul 10x Ammonium Buffer, Tween free, 10,000U
Catalog No. 501655620
Encompass_Preferred
AS ONE Taq DNA Polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa. AS ONE Taq DNA Polymerase has both a 5 prime to 3 prime DNA polymerase and a 5 prime to 3 prime' exonuclease activity. The enzyme lacks a 3 prime to 5 prime exonuclease activity (no proofreading ability). Taq DNA Polymerase leaves an A overhang, which makes the enzyme ideal for TA cloning. Included with Taq is Tween free 10x Ammonium Buffer.
Catalog No. 50-165-5620 Supplier As One International Inc Supplier No. AO111408
AS ONE Taq DNA Polymerase is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa. AS ONE Taq DNA Polymerase has both a 5 prime to 3 prime DNA polymerase and a 5 prime to 3 prime' exonuclease activity. The enzyme lacks a 3 prime to 5 prime exonuclease activity (no proofreading ability). Taq DNA Polymerase leaves an A overhang, which makes the enzyme ideal for TA cloning. Included with Taq is Tween free 10x Ammonium Buffer.