Learn More
Bulldog Bio Inc TaqDog Hot Start DNA Polymerase with 10x Reaction Buffer ( no dNTP), 50,000 Units
SDP
Catalog No. NC1933435
Encompass_Preferred
TaqDog Premium is a great choice for your everyday PCR needs. This recombinant DNA polymerase is derived from Thermus aquaticus is perfect for 3-step, 2-step, and touchdown protocols, as well as completely compatible with common SYBR™ Green-based qPCR buffers. Each kit includes 10x reaction buffer and is optimized for improved amplification efficiency. Better efficiency allows many protocols to be completed in under 1 hour — that is if your thermal cycler can keep up! TaqDog Premium is also a great enzyme to consider for dramatically reducing the cost of your qPCR experiments. Starting at only a few pennies per unit, its ideal for labs performing mouse tail genotyping, and other high volume PCR processing. The purity of TaqDog Premium is so good, that it’s still 100% fully active after 14 days at 37°C. That’s why we ship it at room temperature — a cost savings for your lab and waste savings for the environment.
Catalog No. NC1933435 Supplier Bulldog Bio Inc Supplier No. HTQDG500
TaqDog Premium is a great choice for your everyday PCR needs. This recombinant DNA polymerase is derived from Thermus aquaticus is perfect for 3-step, 2-step, and touchdown protocols, as well as completely compatible with common SYBR™ Green-based qPCR buffers. Each kit includes 10x reaction buffer and is optimized for improved amplification efficiency. Better efficiency allows many protocols to be completed in under 1 hour — that is if your thermal cycler can keep up! TaqDog Premium is also a great enzyme to consider for dramatically reducing the cost of your qPCR experiments. Starting at only a few pennies per unit, its ideal for labs performing mouse tail genotyping, and other high volume PCR processing. The purity of TaqDog Premium is so good, that it’s still 100% fully active after 14 days at 37°C. That’s why we ship it at room temperature — a cost savings for your lab and waste savings for the environment.