Learn More
New England Biolabs, Inc. TaqI-v2 – 20000 units
SDP
Supplier:  New England Biolabs, Inc. R0149M
Encompass
A restriction endonuclease that recognizes the sequence T^CG_A.
Catalog No. 50-812-032
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product