Learn More
New England Biolabs, Inc. TaqI-v2 – 20000 units
SDP
Catalog No. 50812031
Encompass
A restriction endonuclease that recognizes the sequence T^CG_A.
Catalog No. 50-812-031 Supplier New England Biolabs, Inc. Supplier No. R0149L
A restriction endonuclease that recognizes the sequence T^CG_A.