Learn More
New England Biolabs, Inc. TaqI-v2 – 4000 units
SDP
Catalog No. 50812033
Encompass
A restriction endonuclease that recognizes the sequence T^CG_A.
Catalog No. 50-812-033 Supplier New England Biolabs, Inc. Supplier No. R0149S
A restriction endonuclease that recognizes the sequence T^CG_A.