Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Array Human Endogenous Control Panel

Catalog No. 4367563
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 cards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-675-63 2 cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-675-63 Supplier Applied Biosystems™ Supplier No. 4367563
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TaqMan™ Array Human Endogenous Control Panel

The Applied Biosystems™ TaqMan™ Endogenous Control Array is a 384-well micro fluidic card containing 16 human TaqMan™ Gene Expression Assays commonly used as endogenous controls for for commonly used housekeeping genes and genes that exhibit minimal differential expression across different tissues.

TaqMan™ Endogenous Control Array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type Human Endogenous Control Array
Format 384-well Microfluidic Card
Species Human
Content And Storage TaqMan™ Human Endogenous Control Array is sold in a pack of 2 cards and include a CD with assay information and an instrument software input file. The cards should be stored at 4°C.
Form Dried Down
Includes Control Primer-Probe Set(s)
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 2 cards
Reaction Speed Standard
Target Gene Housekeeping & Control Gene Panel
Show More Show Less
What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Can poly A+ purified RNA samples be used with the TaqMan Human Endogenous Control Plate (Cta. No. 4309199)?

No. The plate has been designed to evaluate only total RNA. Ribosomal RNA will be removed from poly A+ purified RNA samples, and is required for accurate detection of the 18s rRNA endogenous control assay used on the plate.

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.