Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Array Human Osteogenesis

Catalog No. 4414096
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 standard plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-140-96 1 standard plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-140-96 Supplier Applied Biosystems™ Supplier No. 4414096
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

TaqMan™ Array Human Osteogenesis

The Applied Biosystems™ TaqMan™ Array Human Osteogenesis 96-well Plate contains 92 assays to osteogenesis associated genes and 4 assays to candidate endogenous control genes.

The panel of assays in this plate targets genes associated with osteoblast determination and differentiation, including the RUNX2 transcription factor and growth factors with induce osteoprogenetor to differentiation and which guide osteoblast function (bone morphogenetic proteins, etc). The panel also includes genes involved in the physiological processes of bone and tooth formation, mineralization and maintenance, including collagens, enamelin and matrix metalloproteinases Gene Signature Plates are 96-well plates that are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.

  • Affordable; minimum order of three plates; ideal for small, medium or large-sized projects
  • Fast delivery; on your bench in days
  • Convenient; dried PCR primer and TaqMan™ probe sets supplied in a 96- well plate; just add master mix and your sample
  • Same TaqMan™Assay Quality, New Format

    TaqMan™ Array Plates contain the same high quality TaqMan™ Gene Expression Assays (PCR primers and TaqMan™ probe sets) that have been commercially available for years in a convenient, dried, 96-well format. Extensive tests have found no difference in performance between TaqMan™Assays supplied in the traditional liquid format and assays dried into a TaqMan™ Array Plate. Figure 1 shows the results of an experiment comparing the linear range of the two formats, where no difference in performance was observed.

    Just Add Master Mix and Sample, and Begin Cycling
    Ideal for projects such as validation of microarray and RNAi data, and pathway studies, the plates include just the right amount of TaqMan™Assay in each well: enough for one 20μl reaction. You can order as few as three Gene Signature Plates, making these an affordable option for many laboratories and minimizing the waste of unused individually purchased TaqMan™Assays. Save a significant amount of time setting up your experiments. Just add master mix and sample, and begin cycling.

    Figure 2 is an example Plate Layout with Gene Symbols and Assay IDs

    Plate Map Ensures Proper Data Labeling and Easy Re-Ordering
    Each pack of TaqMan™ Array Plates is shipped with a CD containing the plate configuration map (see Figure 2) along with the details of each TaqMan™Assay. This information is imported into the real-time PCR instrument to enable proper labeling of experimental data and easy access to Assay ID numbers for simple reordering.

    Simple to Use
    Each TaqMan™ Array Plate well contains a single TaqMan™Assay for one 20μl reaction. The plates can be stored at room temperature or at 4°C. When you are ready to run your assays, simply add PCR master mix and your cDNA sample, seal the plate, and begin cycling on a real-time PCR instrument.

    Compatibility
    TaqMan™ Array Plates have been optimized for use with TaqMan™ Gene Expression Master Mix, but are also compatible with TaqMan™ Universal PCR Master Mix. For studies using cultured cells, TaqMan™ Array Plates are also compatible (and recommended) with the TaqMan™ Gene Expression Cells-to-Ct™ Kit. TaqMan™ Array Plates must be run on a real-time PCR instrument with 96-well plate capability. The plates have been validated for use on Applied Biosystems™ 7000, 7300, 7500 and 7900HT Fast Real-Time PCR Systems using standard 96-well blocks, standard PCR cycling conditions, and 20μl reaction
  • TRUSTED_SUSTAINABILITY

    Specifications

    Type Array Plate
    Format 96-array Plate
    Species Human
    Content And Storage Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.
    Form Dried Down
    Includes TaqMan Assay for One 20 uL Reaction
    Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
    Label or Dye FAM
    Product Line TaqMan
    Purification Method Standard
    Quantity 1 standard plate
    Reaction Speed Standard
    Research Category Cell Cycle & Proliferation, Development, Biomarker-Related Pathway(s), Oncology, Extracellular Matrix & Cell Adhesion
    Show More Show Less
    What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

    The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
    • Inventoried TaqMan array plates: 1 plate
    • TaqMan Flexible Content panel array plates: 6 plates
    • Custom TaqMan array plates: 6 plates
    • Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

    Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

    We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

    Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
    • Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
    • Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

    How do I obtain TaqMan documentation files for my TaqMan array plates?

    Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
    • Select the appropriate product type.
    • Enter the information being requested for the product type.
    • Fixed TaqMan array plates: Enter the Part number and Batch number
    • TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
    • Click Submit.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.