Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Array, Human p53-Mediated Apoptosis, Fast 96-well

Catalog No. 4418812
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96-well plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-188-12 1 x 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-188-12 Supplier Applied Biosystems™ Supplier No. 4418812
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment.

  • These 96-well plates are pre-configured with the most appropriate TaqMan™ Gene Expression Assays for a specific biological process, pathway, or disease state
  • Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment
  • The TaqMan™ Array Human p53 Mediated Apoptosis 96-well Plate contains:
    • 92 assays to genes associated with p53 mediation of apoptosis
    • 4 assays to candidate endogenous control genes
TRUSTED_SUSTAINABILITY

Specifications

Type Array Plate
Format 96-array Plate
Species Human
Content And Storage Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 10 μL reaction. The plates can be stored at room temperature or at 4°C.
Form Dried Down
Includes TaqMan Assay for One 10 uL Reaction
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 1 x 96-well plate
Reaction Speed Fast
Research Category Cell Cycle & Proliferation, Apoptosis, Extracellular Matrix & Cell Adhesion, Immunology, Neurobiology
Show More Show Less
What mastermix can I use for running samples on TaqMan Gene Signature Array Plates?

We recommend that for standard (20 µL) TaqMan Array Plates, you use either the Universal Master Mix (I or II) or the Gene Expression MasterMix. For fast (10 µL) TaqMan Array Plates, use the Fast Universal Master Mix. The Fast Advanced Master Mix can also be used for either plate type, but we recommend following the protocol in this manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025706_TaqManFastAdvMM_UG.pdf).

What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.