Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ TaqMan™ Array Mouse Alzheimer's Panel

Catalog No. 4378714
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 cards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-787-14 4 cards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-787-14 Supplier Applied Biosystems™ Supplier No. 4378714
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TaqMan™ Array Mouse Alzheimer's Panel

The Applied Biosystems™ TaqMan™ Mouse Alzheimer's Array is a 384-well micro flluidic card containing 96 human TaqMan™ Gene Expression Assays. This product is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System and sold in packs of 4.

TaqMan™ Arrays for Alzheimer’s for mouse closely follow the human panel and are from orthologous genes of the human assays. The mouse panel has 92 assays and four controls (18S, actb, hprt1 and ipo8).The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.
TRUSTED_SUSTAINABILITY

Specifications

Type Array Mouse Alzheimer's Panel
Format 384-well Microfluidic Card
Species Mouse
Content And Storage Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.
Form Dried Down
Includes TaqMan Assay for One 2 uL Reaction
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line TaqMan
Quantity 4 cards
Reaction Speed Standard
Research Category Neurobiology
Target Gene 94 assays for APP processing, cholesterol biosynthesis, and other Alzheimer's targets
Show More Show Less
What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.