Learn More
Invitrogen™ TaqMan™ Cells-to-CT™ Express 1-Step Kit
Description
Skip extraction entirely. Go from cell lysate to results in 1 step.
The TaqMan™ Cells-to-CT™ Express 1-Step Kit enables gene expression analysis directly from cultured cell lysates in a single RT-qPCR reaction. By eliminating RNA purification and combining reverse transcription and quantitative PCR into one streamlined workflow, researchers can generate high-quality gene expression data with fewer processing steps and less hands-on time. The workflow is well suited for early-stage research applications, including target identification, target validation, pathway analysis, and high-throughput screening.
Designed for speed and agility, the workflow combines rapid 5-minute cell lysis with integrated 1-step RT-qPCR, allowing researchers to move from cells to gene expression results in approximately 70 minutes.
Simplify RT-qPCR Workflows for Early Target Validation Research
RT-qPCR remains one of the most widely used methods for measuring gene expression during early-stage drug discovery and biological research. However, traditional workflows typically require RNA purification before amplification, adding time, consumables, and multiple handling steps to the process.
Direct-lysis workflows offer an alternative by enabling cells to be processed and analyzed without RNA extraction. When paired with 1-step RT-qPCR, researchers can move seamlessly from sample preparation to gene expression analysis in a single, continuous workflow.
The TaqMan™ Cells-to-CT™ Express 1-Step Kit was developed to support laboratories performing gene expression-based screening and target validation studies, helping reduce workflow complexity while maintaining the sensitivity, reproducibility, and performance required for confident decision-making.
Key Features & Benefits
- Simplified Gene Expression Workflow – Go directly from cultured mammalian cells to RT-qPCR with no RNA purification required and no separate reverse transcription reaction required. Rapid 5-minute cell lysis with no stop solution or heat inactivation step further streamlines sample preparation.
- Reduced Sample Handling – Minimize pipetting and sample transfer steps to reduce opportunities for user error, contamination, and sample loss.
- Lower Plastic and Consumable Usage – Reduce the number of tubes, plates, pipette tips, and reagents required compared to traditional extraction-based workflows.
- Reliable Gene Expression Results – Generate sensitive and reproducible gene expression data directly from cultured cell lysates using trusted TaqMan™ Gene Expression Assays.
- Scalable for High-Throughput Workflows – Compatible with both manual and automated laboratory environments, featuring an automation-ready workflow that enables efficient processing of large numbers of samples.
- Designed for Early Discovery Research – Enable gene expression analysis for target identification, target validation, and screening applications.
Applications
The TaqMan™ Cells-to-CT™Express 1-Step Kit is designed for RT-qPCR-based gene expression workflows in research and early drug discovery screening labs.
Applications include:
- RT-qPCR gene expression analysis
- Target identification and gene expression-based target validation
- Early-stage drug discovery screening
- High-throughput compound screening
- Pathway activation and response studies
- RNA analysis from cultured mammalian cells
- Candidate target prioritization
- Biomarker and gene expression profiling
- RNA-mediated gene regulation studies
Specifications
Specifications
| Format | Kit |
| Isolation Technology | Direct Cell Lysis |
| Sample Type | Mammalian cells |
| Product Type | Direct Cell Lysis RT-qPCR |
| Quantity | 2500 Reactions |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.