Learn More
LAMDA BIOTECH INC TaqProbe Direct-qPCR
Catalog No. 502045788
Encompass_Preferred
TaqProbe Direct-qPCR Kit contains all the reagents needed for quick preparation of genomic DNA and the qPCR master mix for TaqMan real-time PCR assay. Any type of tissues can be used for this kit, such as mouse tails and nail tips, and other animal tissues and cells; plant leaf, root or seeds; bacteria, fungi and other samples
Catalog No. 50-204-5788 Supplier LAMDA BIOTECH INC Supplier No. D501P
TaqProbe Direct-qPCR Kit contains all the reagents needed for quick preparation of genomic DNA and the qPCR master mix for TaqMan real-time PCR assay. Any type of tissues can be used for this kit, such as mouse tails and nail tips, and other animal tissues and cells; plant leaf, root or seeds; bacteria, fungi and other samples