Learn More
Cusabio Technology LLC TAR DNA binding protein
Catalog No. 5000049966
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Human TAR DNA-binding protein 43 (TARDBP/TDP43) in serum plasma cell culture supernates and cerebrospinal fluid (CSF) samples Utilizing a sensitive sandwich assay format the kit demonstrates a broad detection range of 0 312 ng/mL to 20 ng/mL with a low minimum detectable concentration of 0 078 ng/mL ensuring precise measurement across biological matrices The protocol requires 50-100 L of sample per test and delivers results within 1-5 hours through absorbance measurement at 450 nm This kit detects TARDBP with high specificity and reproducibility making it suitable for research applications involving neurological studies biomarker analysis or cellular response monitoring where accurate quantification of TDP-43 protein levels is critical
Catalog No. 50-000-49966 Supplier Cusabio Technology LLC Supplier No. CSBE17007H
This ELISA kit is designed for the quantitative detection of Human TAR DNA-binding protein 43 (TARDBP/TDP43) in serum plasma cell culture supernates and cerebrospinal fluid (CSF) samples Utilizing a sensitive sandwich assay format the kit demonstrates a broad detection range of 0 312 ng/mL to 20 ng/mL with a low minimum detectable concentration of 0 078 ng/mL ensuring precise measurement across biological matrices The protocol requires 50-100 L of sample per test and delivers results within 1-5 hours through absorbance measurement at 450 nm This kit detects TARDBP with high specificity and reproducibility making it suitable for research applications involving neurological studies biomarker analysis or cellular response monitoring where accurate quantification of TDP-43 protein levels is critical