Learn More
Medchemexpress LLC Tarafenacin D-tartrate | 1159101-48-0 | 99.9% | 558.48 | 5 MG
SDP
Catalog No. 5000359778
Encompass_Preferred
Tarafenacin D-tartrate is a highly selective M3 muscarinic receptor antagonist with a Ki of 0.19 nM, demonstrating approximately 200-fold selectivity over the M2 receptor. It is recognized as a potent anti-cholinergic agent for inhibiting carbachol-induced bladder contractions.
  • Highly selective M3 muscarinic receptor antagonist (Ki = 0.19 nM)
  • Exhibits 200-fold selectivity over M2 receptor
  • Potent anti-cholinergic agent for inhibiting carbachol-induced bladder contractions
  • High urinary versus cardiac selectivity (199-fold)
  • High binding affinity (Kd = 0.4 nM) to M5 mAChR
  • Inhibits spontaneous bladder contractions at low doses
  • Investigated in clinical trials for overactive bladder (OAB)
Catalog No. 50-003-59778 Supplier Medchemexpress LLC Supplier No. HY14825A5MG
Tarafenacin D-tartrate is a highly selective M3 muscarinic receptor antagonist with a Ki of 0.19 nM, demonstrating approximately 200-fold selectivity over the M2 receptor. It is recognized as a potent anti-cholinergic agent for inhibiting carbachol-induced bladder contractions.
  • Highly selective M3 muscarinic receptor antagonist (Ki = 0.19 nM)
  • Exhibits 200-fold selectivity over M2 receptor
  • Potent anti-cholinergic agent for inhibiting carbachol-induced bladder contractions
  • High urinary versus cardiac selectivity (199-fold)
  • High binding affinity (Kd = 0.4 nM) to M5 mAChR
  • Inhibits spontaneous bladder contractions at low doses
  • Investigated in clinical trials for overactive bladder (OAB)