Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taranabant (1R,2R) stereoisomer | 701977-08-4 | 98.2% | 515.95 | C27H25ClF3N3O2 | 5 MG
SDP

Catalog No. 5000215880
Encompass_Preferred

Taranabant ((1R,2R) stereoisomer) is the R-enantiomer of taranabant, a potent and selective cannabinoid-1 (CB1) receptor inverse agonist used for research. The material is supplied as a white to off-white solid and includes documented purity, molecular weight, solubility, and recommended storage conditions for laboratory use.

  • High purity: 98.15%.
  • Molecular weight: 515.95 g/mol.
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic agitation).
  • Stable as powder at -20°C for long-term storage.
  • Supplied with supporting datasheet specifications for research handling.

Catalog No. 50-002-15880 Supplier Medchemexpress LLC Supplier No. HY10013B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Taranabant ((1R,2R) stereoisomer) is the R-enantiomer of taranabant, a potent and selective cannabinoid-1 (CB1) receptor inverse agonist used for research. The material is supplied as a white to off-white solid and includes documented purity, molecular weight, solubility, and recommended storage conditions for laboratory use.

  • High purity: 98.15%.
  • Molecular weight: 515.95 g/mol.
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic agitation).
  • Stable as powder at -20°C for long-term storage.
  • Supplied with supporting datasheet specifications for research handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.