Learn More
Medchemexpress LLC HY-10013 25mg , Taranabant MK-0364 CAS:701977-09-5 Purity:98%
SDP
Catalog No. 501493993
Encompass_Preferred
Medchemexpress, HY-10013 25mg Taranabant MK-0364 CAS:701977-09-5 Formula:C27H25ClF3N3O2 IC50: 0.3 nM (hCB1R), 0.4 nM (rCB1R) Ki: 0.13 nM (hCB1R), 0.27 nM (rCB1R) Purity:98% Taranabant is a highly potent and selective cannabinoid 1 ( CB1 ) receptor inverse agonist that inhibits the binding and functional activity of various agonists, with a binding K i of 0.13 nM for the human CB1R in vitro. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3993 Supplier Medchemexpress LLC Supplier No. HY1001325MG
Medchemexpress, HY-10013 25mg Taranabant MK-0364 CAS:701977-09-5 Formula:C27H25ClF3N3O2 IC50: 0.3 nM (hCB1R), 0.4 nM (rCB1R) Ki: 0.13 nM (hCB1R), 0.27 nM (rCB1R) Purity:98% Taranabant is a highly potent and selective cannabinoid 1 ( CB1 ) receptor inverse agonist that inhibits the binding and functional activity of various agonists, with a binding K i of 0.13 nM for the human CB1R in vitro. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.