Learn More
Medchemexpress LLC HY-10013 2mg Medchemexpress, Taranabant CAS:701977-09-5 Purity:>98%
SDP
Catalog No. 501931724
Encompass_Preferred
Medchemexpress, HY-10013 2mg Taranabant CAS:701977-09-5 Taranabant is a highly potent and selective cannabinoid 1 ( CB1 ) receptor inverse agonist that inhibits the binding and functional activity of various agonists, with a binding K i of 0.13 nM for the human CB1R in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1724 Supplier Medchemexpress LLC Supplier No. HY100132MG
Medchemexpress, HY-10013 2mg Taranabant CAS:701977-09-5 Taranabant is a highly potent and selective cannabinoid 1 ( CB1 ) receptor inverse agonist that inhibits the binding and functional activity of various agonists, with a binding K i of 0.13 nM for the human CB1R in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.