Learn More
Medchemexpress LLC Taranabant | 701977-08-4 | MFCD30534391 | 98.2% | 515.95 | C27H25ClF3N3O2 | 25 MG
SDP
Catalog No. 5000215834
Encompass_Preferred
Taranabant ((1R,2R)stereoisomer) is the R-enantiomer of taranabant, a potent and selective cannabinoid 1 (CB1) receptor inverse agonist intended for research use. The material is supplied as a solid or as DMSO solutions and is characterized for purity and identity for biochemical and cellular studies.
  • R-enantiomer of taranabant, enabling stereospecific studies.
  • Selective CB1 receptor inverse agonist for receptor pharmacology assays.
  • High purity suitable for biochemical and cellular experiments.
  • Available as solid and as DMSO solutions for flexible dosing and formulation.
  • Characterized with CAS number 701977-08-4 and molecular weight 515.95.
Catalog No. 50-002-15834 Supplier Medchemexpress LLC Supplier No. HY10013B25MG
Taranabant ((1R,2R)stereoisomer) is the R-enantiomer of taranabant, a potent and selective cannabinoid 1 (CB1) receptor inverse agonist intended for research use. The material is supplied as a solid or as DMSO solutions and is characterized for purity and identity for biochemical and cellular studies.
  • R-enantiomer of taranabant, enabling stereospecific studies.
  • Selective CB1 receptor inverse agonist for receptor pharmacology assays.
  • High purity suitable for biochemical and cellular experiments.
  • Available as solid and as DMSO solutions for flexible dosing and formulation.
  • Characterized with CAS number 701977-08-4 and molecular weight 515.95.