Learn More
Medchemexpress LLC HY-10013A 5mg Medchemexpress, Taranabant (racemate) CAS:701977-00-6 Purity:>98%
SDP
Catalog No. 502080577
Encompass_Preferred
Medchemexpress, HY-10013A 5mg Taranabant (racemate) CAS:701977-00-6 Taranabant racemate (MK-0364 racemate) is an antagonist and/or inverse agonist of the Cannabinoid-1 (CB1) receptor extracted from patent WO 2004048317 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0577 Supplier Medchemexpress LLC Supplier No. HY10013A5MG
Medchemexpress, HY-10013A 5mg Taranabant (racemate) CAS:701977-00-6 Taranabant racemate (MK-0364 racemate) is an antagonist and/or inverse agonist of the Cannabinoid-1 (CB1) receptor extracted from patent WO 2004048317 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.