Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taranabant racemate (MK-0364 racemate) | 701977-00-6 | 99.8% | 515.95 | C27H25ClF3N3O2 | 100 MG
SDP

Catalog No. 5000215564
Encompass_Preferred

Taranabant racemate (MK-0364 racemate) is a research-grade small-molecule antagonist and/or inverse agonist of the cannabinoid-1 (CB1) receptor. Supplied as a high-purity solid, it is intended for in vitro and preclinical research; consult the manufacturer's datasheet and SDS for handling, storage, and safety information.

  • Acts as a cannabinoid-1 (CB1) receptor antagonist and/or inverse agonist.
  • High purity (99.84% as documented by the manufacturer).
  • Molecular formula C27H25ClF3N3O2.
  • Molecular weight 515.95.
  • Listed CAS number 701977-00-6.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-15564 Supplier Medchemexpress LLC Supplier No. HY10013A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Taranabant racemate (MK-0364 racemate) is a research-grade small-molecule antagonist and/or inverse agonist of the cannabinoid-1 (CB1) receptor. Supplied as a high-purity solid, it is intended for in vitro and preclinical research; consult the manufacturer's datasheet and SDS for handling, storage, and safety information.

  • Acts as a cannabinoid-1 (CB1) receptor antagonist and/or inverse agonist.
  • High purity (99.84% as documented by the manufacturer).
  • Molecular formula C27H25ClF3N3O2.
  • Molecular weight 515.95.
  • Listed CAS number 701977-00-6.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.