Learn More
Sigma Aldrich Fine Chemicals Biosciences Taraxasterol acetate phypr10MG
Catalog No. 502902648
Encompass
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG
Catalog No. 50-290-2648 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHL8054210MG
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG