Learn More
Medchemexpress LLC Taraxerol | 127-22-0 | MFCD13186859 | 98.0% | 426.72 g·mol⁻¹ | C30H50O | 10 MG
SDP
Catalog No. 5000289472
Encompass_Preferred
Taraxerol is a naturally occurring pentacyclic triterpenoid reported to have anti-inflammatory and anticancer activity. It attenuates acute inflammation by inhibiting NF-κB signaling and can induce apoptosis in cellular models. Supplied as a high-purity research reagent for laboratory use.
  • Natural pentacyclic triterpenoid with anti-inflammatory and anticancer activity.
  • Inhibits NF-κB signaling and can induce apoptosis in cells.
  • High purity (typical 98.0%; batch COA may report higher).
  • Supplied in small research pack sizes suitable for biochemical and cell studies.
  • Soluble in common organic solvents such as DMSO and THF.
  • Intended for laboratory research use only.
Catalog No. 50-002-89472 Supplier Medchemexpress LLC Supplier No. HYN247710MG
Taraxerol is a naturally occurring pentacyclic triterpenoid reported to have anti-inflammatory and anticancer activity. It attenuates acute inflammation by inhibiting NF-κB signaling and can induce apoptosis in cellular models. Supplied as a high-purity research reagent for laboratory use.
  • Natural pentacyclic triterpenoid with anti-inflammatory and anticancer activity.
  • Inhibits NF-κB signaling and can induce apoptosis in cells.
  • High purity (typical 98.0%; batch COA may report higher).
  • Supplied in small research pack sizes suitable for biochemical and cell studies.
  • Soluble in common organic solvents such as DMSO and THF.
  • Intended for laboratory research use only.