Learn More
Aviva Systems Biology Corp TARDBP Antibody - middle region (ARP35837_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35837_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TARDBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TARDBP. Alternate names: TDP-43, ALS10ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55885