Learn More
Medchemexpress LLC HY-10291 100mg Medchemexpress, Tarenflurbil CAS:51543-40-9 Purity:>98%
SDP
Catalog No. 502080654
Encompass_Preferred
Medchemexpress, HY-10291 100mg Tarenflurbil CAS:51543-40-9 Tarenflurbil ((R)-Flurbiprofen) is the R-enantiomer of the racemate NSAID Flurbiprofen, Tarenflurbil ((R)-Flurbiprofen) inhibits the binding of [3H]9-cis-RA to RXRα LBD with IC50 of 75 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0654 Supplier Medchemexpress LLC Supplier No. HY10291100MG
Medchemexpress, HY-10291 100mg Tarenflurbil CAS:51543-40-9 Tarenflurbil ((R)-Flurbiprofen) is the R-enantiomer of the racemate NSAID Flurbiprofen, Tarenflurbil ((R)-Flurbiprofen) inhibits the binding of [3H]9-cis-RA to RXRα LBD with IC50 of 75 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.